Anti-IL12 p40 Goat Polyclonal Antibody
Catalog # BIRBORB19735-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen symbol:IL12 p40
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Rat,Mouse
- Western blot:Yes
- Storage buffer:Supplied at 0,5 mg/ml in Tris saline, 0,02% sodium azide, pH 7,3 with 0,5% BSA.
- Sequence:(C)EARQQNKYENYS
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:C-EARQQNKYENYS
- Purification:purified by ammonium sulphate precipitation followed by antigen affinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: IL12 p40
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Mouse, Rat