Order Entry
Austria
ContactUsLinkComponent
 
Anti-PI3-kinase p85-alpha/ gamma Rabbit Polyclonal Antibody
 :  Biorbyt
 
undefined
Anti-PI3-kinase p85-alpha/ gamma Rabbit Polyclonal Antibody
 :  Biorbyt

 

  • Catalog No:
  • BIRBORB6714-100
  • BIRBORB6714-20
  • Antigen name:
  • PI3-kinase p85-alpha/ gamma
  • PI3-kinase p85-alpha/ gamma
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human, Rat, Mouse
  • Human, Rat, Mouse
  • Host:
  • Rabbit
  • Rabbit
  • Isotype:
  • IgG
  • IgG
  • Western blot:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Modification:
  • phospho-Tyr467, phospho-Tyr199
  • phospho-Tyr467, phospho-Tyr199
  • Immunogen:
  • A synthesized peptide derived from human PI3-kinase p85- alpha/ gamma around the phosphorylation site of Tyrosine 467/199
  • A synthesized peptide derived from human PI3-kinase p85- alpha/ gamma around the phosphorylation site of Tyrosine 467/199
  • Molecular weight:
  • 54.83 kDa
  • 54.83 kDa
  • Purification:
  • purified by immunoaffinity chromatography
  • purified by immunoaffinity chromatography
  • Concentration:
  • 20 ug: 0,2 mg/ml; 50 ug: 0,5 mg/ml; 100 ug: 1 ug/ul; 200 ug: 1 ug/ul
  • 20 ug: 0,2 mg/ml; 50 ug: 0,5 mg/ml; 100 ug: 1 ug/ul; 200 ug: 1 ug/ul
  • Storage buffer:
  • Phosphate buffer saline, pH. 7.4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
  • Phosphate buffer saline, pH. 7.4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

 

 

Frequently Bought Together