Anti-MAPK14 Rabbit Polyclonal Antibody
: Biorbyt
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: MAPK14 (mitogen-activated protein kinase 14)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
- Catalog No:
- BIRBORB6578-20
- BIRBORB6578-5
- BIRBORB6578-100
- Antigen symbol:
- MAPK14
- MAPK14
- MAPK14
- Antigen name:
- Mitogen-activated protein kinase 14
- Mitogen-activated protein kinase 14
- Mitogen-activated protein kinase 14
- Antibody type:
- Primary
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Host:
- Rabbit
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- IgG
- Western blot:
- Yes
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Yes
- Modification:
- phospho-Thr180, phospho-Tyr182
- phospho-Thr180, phospho-Tyr182
- phospho-Thr180, phospho-Tyr182
- Immunogen:
- KLH conjugated synthetic phosphopeptide derived from human p38 MAPK around the phosphorylation site of(Thr180/Tyr182).
- KLH conjugated synthetic phosphopeptide derived from human p38 MAPK around the phosphorylation site of(Thr180/Tyr182).
- KLH conjugated synthetic phosphopeptide derived from human p38 MAPK around the phosphorylation site of(Thr180/Tyr182).
- Form:
- liquid
- liquid
- liquid
- Molecular weight:
- 41 kDa
- 41 kDa
- 41 kDa
- Purification:
- purified by affinity chromatography
- purified by affinity chromatography
- purified by affinity chromatography
- Concentration:
- 0,20 mg/ml
- 0,05 mg/ml
- 0,5 mg/ml
- Storage buffer:
- 10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- 10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- 10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- Storage temperature:
- Cool
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
- Ice
Frequently Bought Together