Anti-Metronidazole (1C2) Mouse monoclonal Antibody [clone: 1C2] FITC (Fluorescein)
Catalog # BIRBORB4287-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Metronidazole
- Clonality:Monoclonal
- Clone:1C2
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG
- Form:liquid
- Storage buffer:10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- Storage temperature:Cool
- Concentration:0,5 mg/ml
- Shipping temperature:Ice
- Immunogen:KLH conjugated synthetic peptide derived from human Ceruloplasmin.
- Purification:purified by affinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)
Type: Primary
Antigen: Metronidazole (1C2)
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: