Anti-CERS6 Mouse Monoclonal Antibody [clone: 5H7]
Catalog # ABNOH00253782-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Ceramide synthase 6
- Antigen symbol:CERS6
- Clonality:Monoclonal
- Clone:5H7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:253782
- Antigen synonyms:CERS5|LASS6
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: CERS6
Clonality: Monoclonal
Clone: 5H7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human