Order Entry
Austria
ContactUsLinkComponent
Anti-E2F8 Mouse Monoclonal Antibody [clone: 3E9-2F5]
Anti-E2F8 Mouse Monoclonal Antibody [clone: 3E9-2F5]
Catalog # ABNOH00079733-M01
Supplier:  Abnova
undefined
Anti-E2F8 Mouse Monoclonal Antibody [clone: 3E9-2F5]
Catalog # ABNOH00079733-M01
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    E2F transcription factor 8
  • Antigen symbol:
    E2F8
  • Clonality:
    Monoclonal
  • Clone:
    3E9-2F5
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Gene ID:
    79733
  • Antigen synonyms:
    E2F-8
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Type: Primary
Antigen: E2F8
Clonality: Monoclonal
Clone: 3E9-2F5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human