Anti-GSDMB Mouse Monoclonal Antibody [clone: 1E10]
Catalog # ABNOH00055876-M05
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Gasdermin B
- Antigen symbol:GSDMB
- Clonality:Monoclonal
- Clone:1E10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:55876
- Antigen synonyms:PRO2521|PP4052|GSDML
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: GSDMB
Clonality: Monoclonal
Clone: 1E10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human