Anti-DHTKD1 Mouse Polyclonal Antibody
Catalog # ABNOH00055526-B01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:dehydrogenase E1 And transketolase Domain Containing 1
- Antigen symbol:DHTKD1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:55526
- Antigen synonyms:CMT2Q|AMOXAD
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Specifications
About this item
Type: Primary
Antigen: DHTKD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human