Anti-ZWINT Rabbit Polyclonal Antibody
Catalog # ABNOH00011130-D01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:ZW10 interacting kinetochore protein
- Antigen symbol:ZWINT
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Cross adsorption:No
- Gene ID:11130
- Antigen synonyms:ZWINT1|human ZW10 interacting protein-1|zwint-1|KNTC2AP|ZW10-interacting protein 1|ZW10 interactor|HZwint-1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: ZWINT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human