Anti-ARPC3 Mouse Monoclonal Antibody [clone: 2E11]
ABNOH00010094-M06
: Abnova
- Antibody type:Primary
- Antigen name:Actin related protein 2/3 complex, subunit 3, 21kDa
- Antigen symbol:ARPC3
- Clonality:Monoclonal
- Clone:2E11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:10094
- Antigen synonyms:Arp2/3 complex 21 kDa subunit|Actin-related protein 2/3 complex subunit 3|ARPC3|ARC21|p21-ARC
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: ARPC3
Clonality: Monoclonal
Clone: 2E11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human