Anti-TFF1 Mouse Monoclonal Antibody [clone: M1]
Catalog # ABNOH00007031-M03
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Trefoil factor 1
- Antigen symbol:TFF1
- Clonality:Monoclonal
- Clone:M1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:7031
- Antigen synonyms:TFF 1|HP1.A|Breast cancer estrogen-inducible protein|pS2 protein|TFF-1|TFF1|trefoil factor 1|PNR-2|trefoil factor
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: TFF1
Clonality: Monoclonal
Clone: M1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human