Order Entry
Austria
ContactUsLinkComponent
Anti-TCF19 Mouse Monoclonal Antibody [clone: 6D8]
Artikelnr. ABNOH00006941-M01
Lieferant:  Abnova
undefined
Anti-TCF19 Mouse Monoclonal Antibody [clone: 6D8]
Artikelnr. ABNOH00006941-M01
Lieferant:  Abnova

Spezifikationen

  • Antibody type:
    Primary
  • Antigen name:
    Transcription Factor 19
  • Antigen symbol:
    TCF19
  • Clonality:
    Monoclonal
  • Clone:
    6D8
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • ChIP:
    Yes
  • Gene ID:
    6941
  • Antigen synonyms:
    SC1(TCF19)-7|SC1(TCF19)-6|SC1|transcription factor 19|DAMA-213L4.5|SC1-1|SCI(TCF19)-4|transcription factor 19 (SC1)
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

Spezifikationen

Über diesen Artikel

Type: Primary
Antigen: TCF19
Clonality: Monoclonal
Clone: 6D8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human