Order Entry
Austria
ContactUsLinkComponent
Anti-TAF7 Mouse Monoclonal Antibody [clone: 2C5]
Anti-TAF7 Mouse Monoclonal Antibody [clone: 2C5]
Artikelnr. ABNOH00006879-M01
Lieferant:  Abnova
undefined
Anti-TAF7 Mouse Monoclonal Antibody [clone: 2C5]
Artikelnr. ABNOH00006879-M01
Lieferant:  Abnova

Spezifikationen

  • Antibody type:
    Primary
  • Antigen name:
    TAF7 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 55kDa
  • Antigen symbol:
    TAF7
  • Clonality:
    Monoclonal
  • Clone:
    2C5
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • ChIP:
    Yes
  • Gene ID:
    6879
  • Antigen synonyms:
    TAF2F|TAFII55
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

Spezifikationen

Über diesen Artikel

Type: Primary
Antigen: TAF7
Clonality: Monoclonal
Clone: 2C5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human