Anti-PGM3 Mouse Monoclonal Antibody [clone: 1E2-1B12]
Catalog # ABNOH00005238-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Phosphoglucomutase 3
- Antigen symbol:PGM3
- Clonality:Monoclonal
- Clone:1E2-1B12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- ChIP:Yes
- Gene ID:5238
- Antigen synonyms:IMD23|AGM1|PGM 3|PAGM
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: PGM3
Clonality: Monoclonal
Clone: 1E2-1B12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human