Anti-NAB2 Mouse Polyclonal Antibody
ABNOH00004665-B01P
: Abnova
- Antibody type:Primary
- Antigen name:NGFI-A binding Protein 2
- Antigen symbol:NAB2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:4665
- Antigen synonyms:NGFI-A-binding protein 2|NAB2|MADER|EGR-1-binding protein 2|Melanoma-associated delayed early response protein|Protein MADER
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Type: Primary
Antigen: NAB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human