Anti-LYN Mouse Polyclonal Antibody
ABNOH00004067-B01P
: Abnova
- Antibody type:Primary
- Antigen name:Tyrosine-protein kinase Lyn
- Antigen symbol:LYN
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:4067
- Antigen synonyms:LYN proto-oncogene|p53Lyn|p56Lyn|JTK8|Src family tyrosine kinase
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Type: Primary
Antigen: LYN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human