Anti-IL13RA2 Mouse Polyclonal Antibody
Catalog # ABNOH00003598-B01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Interleukin 13 Receptor Alpha 2
- Antigen symbol:IL13RA2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:3598
- Antigen synonyms:CD213A2|IL-13R|CT19|IL13BP
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Specifications
About this item
Type: Primary
Antigen: IL13RA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human