Order Entry
Austria
ContactUsLinkComponent
 
Anti-G6PD Rabbit Polyclonal Antibody
  ABNOH00002539-D01P
 :  Abnova
undefined
Anti-G6PD Rabbit Polyclonal Antibody
  ABNOH00002539-D01P
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Glucose-6-phosphate dehydrogenase
  • Antigen symbol:
    G6PD
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    2539
  • Antigen synonyms:
    G6PD1|glucose-6-phosphate 1-dehydrogenas
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

 

 

Type: Primary
Antigen: G6PD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human