Anti-FKBP5 Mouse Polyclonal Antibody
Catalog # ABNOH00002289-B01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:FK506 binding protein 5
- Antigen symbol:FKBP5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:2289
- Antigen synonyms:AIG6|FKBP51|P54|FKBP54|PPIase|Ptg-10
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Specifications
About this item
Type: Primary
Antigen: FKBP5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human