Anti-ERN1 Mouse Monoclonal Antibody [clone: 8D12]
Catalog # ABNOH00002081-M02
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Endoplasmic reticulum to nucleus signaling 1
- Antigen symbol:ERN1
- Clonality:Monoclonal
- Clone:8D12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:2081
- Antigen synonyms:ER to nucleus signalling 1|inositol-requiring 1|protein kinase/endoribonuclease|endoplasmic reticulum to nucleus signaling 1|IRE1P|MGC163279|endoplasmic reticulum to nucleus signalling 1|IRE1a|IRE1-alpha|MGC163277|ERN1|IRE1|FLJ30999
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: ERN1
Clonality: Monoclonal
Clone: 8D12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human