Order Entry
Austria
ContactUsLinkComponent
Anti-DOCK2 Rabbit Polyclonal Antibody
Anti-DOCK2 Rabbit Polyclonal Antibody
Catalog # ABNOH00001794-D01
Supplier:  Abnova
undefined
Anti-DOCK2 Rabbit Polyclonal Antibody
Catalog # ABNOH00001794-D01
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    dedicator of cytokinesis 2
  • Antigen symbol:
    DOCK2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    1794
  • Antigen synonyms:
    IMD40
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

Type: Primary
Antigen: DOCK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human