Anti-TRIM23 Mouse Monoclonal Antibody [clone: 3E8]
Catalog # ABNOH00000373-M05
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Tripartite Motif Containing 23
- Antigen symbol:TRIM23
- Clonality:Monoclonal
- Clone:3E8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:373
- Antigen synonyms:RNF46|ARFD1|ARD1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: TRIM23
Clonality: Monoclonal
Clone: 3E8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human