Anti-ALOX12 Mouse Polyclonal Antibody
ABNOH00000239-B01P
: Abnova
- Antibody type:Primary
- Antigen name:Arachidonate 12-lipoxygenase
- Antigen symbol:ALOX12
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:239
- Antigen synonyms:12-LOX|LOG12|12S-LOX
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Type: Primary
Antigen: ALOX12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human