Order Entry
Austria
ContactUsLinkComponent
Anti-DOCK2 Mouse Monoclonal Antibody [clone: OTI7G2]
Anti-DOCK2 Mouse Monoclonal Antibody [clone: OTI7G2]
Catalog # ORIGTA802698
Supplier:  OriGene
undefined
Anti-DOCK2 Mouse Monoclonal Antibody [clone: OTI7G2]
Catalog # ORIGTA802698
Supplier:  OriGene

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    dedicator of cytokinesis 2
  • Antigen symbol:
    DOCK2
  • Clonality:
    Monoclonal
  • Clone:
    OTI7G2
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Form:
    Liquid
  • Amino acid number:
    1544 to 1830
  • Storage buffer:
    PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Immunogen:
    Protein
  • Purification:
    purified by affinity chromatography
  • Pk:
    100 µl

Specifications

About this item

Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: DOCK2 (dedicator of cytokinesis 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human