Order Entry
Austria
ContactUsLinkComponent
 
Anti-MEF2C Mouse Monoclonal Antibody [clone: OTI1B6]
Catalog # ORIGTA700065
Supplier:  OriGene
Anti-MEF2C Mouse Monoclonal Antibody [clone: OTI1B6]
Catalog # ORIGTA700065
Supplier:  OriGene

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Myocyte enhancer factor 2C
  • Antigen symbol:
    MEF2C
  • Clonality:
    Monoclonal
  • Clone:
    OTI1B6
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Epitope:
    Full length
  • Cross adsorption:
    none
  • Form:
    Liquid
  • Storage buffer:
    Stored in PBS (pH 7,4) with 0,05% sodium azide, 10mg/ml BSA, 50% glycerol
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    50 µG

Specifications

About this item

Recommended Dilutions: 1:250-1:1000

Type: Primary
Antigen: MEF2C (myocyte enhancer factor 2C)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human