Anti-SIVA1 Mouse Monoclonal Antibody [clone: OTI2D3]
ORIGTA506835
: OriGene
- Antibody type:Primary
- Antigen name:SIVA1, apoptosis-inducing factor
- Antigen symbol:SIVA1
- Clonality:Monoclonal
- Clone:OTI2D3
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:18.5 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Pk:100 µl
Recommended Dilutions: Western Blot: 1:1000
Type: Primary
Antigen: SIVA1 (SIVA1, apoptosis-inducing factor)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human