Anti-ACAA2 Mouse Monoclonal Antibody [clone: OTI2H2]
ORIGTA506155
: OriGene
- Antibody type:Primary
- Antigen name:Acetyl-CoA acyltransferase 2
- Antigen symbol:ACAA2
- Clonality:Monoclonal
- Clone:OTI2H2
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:Full length
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:41.7 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Pk:100 µl
Recommended Dilutions: Western Blot: 1:200-4000; Immunochemistry: 1:150; Immunofluorescence: 1:100
Type: Primary
Antigen: ACAA2 (acetyl-CoA acyltransferase 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: