Anti-GADD45G Mouse Monoclonal Antibody [clone: OTI5G2]
ORIGTA505535
: OriGene
- Antibody type:Primary
- Antigen name:growth arrest and DNA-damage-inducible, gamma
- Antigen symbol:GADD45G
- Clonality:Monoclonal
- Clone:OTI5G2
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2b
- Reactivity:Human
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:16.9 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Pk:100 µl
Recommended Dilutions: Western Blot: 1:2000
Type: Primary
Antigen: GADD45G (growth arrest and DNA-damage-inducible, gamma)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human