Anti-TRMT2A Mouse Monoclonal Antibody [clone: OTI3D8]
ORIGTA505517
: OriGene
- Antibody type:Primary
- Antigen name:tRNA methyltransferase 2 homolog A (S. cerevisiae)
- Antigen symbol:TRMT2A
- Clonality:Monoclonal
- Clone:OTI3D8
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:68.5 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Pk:100 µl
Recommended Dilutions: Western Blot: 1:400-4000; Immunochemistry: 1:150; Immunofluorescence: 1:100
Type: Primary
Antigen: TRMT2A (tRNA methyltransferase 2 homolog A (S. cerevisiae))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human