Anti-ATIC Mouse Monoclonal Antibody [clone: OTI2C10]
Catalog # ORIGTA504554
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:5-Aminoimidazole-4-Carboxamide Ribonucleotide Formyltransferase/IMP Cyclohydrolase
- Antigen symbol:ATIC
- Clonality:Monoclonal
- Clone:OTI2C10
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:64.4 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:500-2000
Type: Primary
Antigen: ATIC (5-aminoimidazole-4-carboxamide ribonucleotide formyltransferase/IMP cyclohydrolase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: