Anti-EIF2S1 Mouse Monoclonal Antibody [clone: OTI3H4]
ORIGTA501313
: OriGene
- Antibody type:Primary
- Antigen name:Eukaryotic translation initiation factor 2, subunit 1 alpha, 35kDa
- Antigen symbol:EIF2S1
- Clonality:Monoclonal
- Clone:OTI3H4
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS (pH 7,3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:35.9 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Pk:100 µl
Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100
Type: Primary
Antigen: EIF2S1 (eukaryotic translation initiation factor 2, subunit 1 alpha, 35kDa)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: