Anti-PRPSAP2 Rabbit Polyclonal Antibody
ORIGTA308166
: OriGene
- Antibody type:Primary
- Antigen name:phosphoribosyl pyrophosphate synthetase-associated Protein 2
- Antigen symbol:PRPSAP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Cross adsorption:none
- Form:Liquid
- Amino acid number:1 to 369
- Storage buffer:0,1M Tris, 0,1M Glycine, 10% Glycerol (pH7). 0,01% Thimerosal was added as a preservative.
- Molecular weight:41 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Protein
- Purification:purified by antigen affinity chromatography
- Pk:100 µl
Recommended Dilutions: Immunochemistry/Immunofluorescence:1:100-1:1000; Western Blot:1:500-1:3000
Type: Primary
Antigen: PRPSAP2 (phosphoribosyl pyrophosphate synthetase-associated protein 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse