Anti-RNASEH2A Rabbit Polyclonal Antibody
Catalog # ORIGTA306706
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:Ribonuclease H2, Subunit A
- Antigen symbol:RNASEH2A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS containing 0,02% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography via peptide column
- Pk:100 µG
Specifications
About this item
Recommended Dilutions: Western Blot: 1 ?g/ml; Immunochemistry: 2 ?g/ml; Immunofluorescence: 4 ?g/ml
Type: Primary
Antigen: RNASEH2A (ribonuclease H2, subunit A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat