Anti-CST3 Goat antibody
Catalog # ORIGTA305611
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:Cystatin C
- Antigen symbol:CST3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:internal
- Cross adsorption:none
- Form:Liquid
- Storage buffer:0,5 mg/ml in Tris saline, 0,02% sodium azide, pH 7,3 with 0,5% BSA
- Sequence:C-SVEEEGVRRALD
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide. Supplied at 0.5 mg/ml in Tris saline, 0.02% sodium azide, pH7.3 with 0.5% bovine serum albumin. Aliquot and store at -20C. Minimize freezing and thawing.
- Pk:100 µG
Specifications
About this item
Recommended Dilutions: Western Blot: 03-1?g/ml
Type: Primary
Antigen: CST3 (cystatin C)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human