The A20 antibody reacts with mouse CD45.1, also known as Ly5.1, which is a strain-specific allelic form of the CD45 Leukocyte Common Antigen (LCA). Functionally, CD45 is a protein tyrosine phosphatase whose broad cell distribution supports a critical role in many leukocyte functions, including regulation of signal transduction and cell activation associated with the T cell and B cell receptors.
The A20 antibody is typically used as a leukocyte marker in Ly5.1 mouse strains: SJL/J, DA, STS/A and RIII. The antibody has been demonstrated to specific for CD45.1 and is not cross-reactive with CD45.2 bearing cells.
Type: Primary
Antigen: CD45.1 (CD45.1)
Clonality: monoclonal
Clone: A20
Conjugation: violetFluor® 450
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Mouse
- Catalog No:
- 75-0453-U100
- 75-0453-U025
- Antigen symbol:
- PTPRC
- PTPRC
- Antigen name:
- Protein tyrosine phosphatase receptor type C
- Protein tyrosine phosphatase receptor type C
- Antigen synonyms:
- CD45R|B220|CD45|GP180|T200|L-CA|LY5|receptor type C|LCA|protein tyrosine phosphatase
- CD45R|B220|CD45|GP180|T200|L-CA|LY5|receptor type C|LCA|protein tyrosine phosphatase
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- VioletFluor® 450
- VioletFluor® 450
- Clone:
- A20
- A20
- Reactivity:
- Mouse
- Mouse
- Host:
- Mouse
- Mouse
- Gene ID:
- 19264
- 19264
- Isotype:
- IgG2a kappa
- IgG2a kappa
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Cross adsorption:
- none
- none
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice