Order Entry
Austria
ContactUsLinkComponent
Anti-CD45RO Mouse Monoclonal Antibody (Peridinin Chlorophyll/Cy5.5®) [clone: UCHL1]
Anti-CD45RO Mouse Monoclonal Antibody (Peridinin Chlorophyll/Cy5.5®) [clone: UCHL1]
Supplier:  Tonbo Biosciences
Certificates
undefined
Anti-CD45RO Mouse Monoclonal Antibody (Peridinin Chlorophyll/Cy5.5®) [clone: UCHL1]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 65-0457-T100
  • 65-0457-T025
  • Antigen symbol:
  • PTPRC
  • PTPRC
  • Antigen name:
  • Protein tyrosine phosphatase receptor type C
  • Protein tyrosine phosphatase receptor type C
  • Antigen synonyms:
  • CD45R|B220|CD45|GP180|T200|L-CA|LY5|receptor type C|LCA|protein tyrosine phosphatase
  • CD45R|B220|CD45|GP180|T200|L-CA|LY5|receptor type C|LCA|protein tyrosine phosphatase
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
  • PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
  • Clone:
  • UCHL1
  • UCHL1
  • Reactivity:
  • Chimpanzee, Human
  • Chimpanzee, Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 5788
  • 5788
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Neutralisation:
  • Yes
  • Yes
  • Cross adsorption:
  • none
  • none
  • Storage temperature:
  • Store undiluted at 2...8 °C, protect from light, do not freeze
  • Store undiluted at 2...8 °C, protect from light, do not freeze
  • Shipping temperature:
  • Ice
  • Ice
  • Tested applications:
  • Flow Cytometry
  • Flow Cytometry

Specifications

Related Information

Frequently Bought Together