Order Entry
Austria
ContactUsLinkComponent
Anti-CTLA4 Armenian Hamster Monoclonal Antibody (PE (Phycoerythrin)) [clone: UC10-4F10-11]
Anti-CTLA4 Armenian Hamster Monoclonal Antibody (PE (Phycoerythrin)) [clone: UC10-4F10-11]
Supplier:  Tonbo Biosciences
Certificates
undefined
Anti-CTLA4 Armenian Hamster Monoclonal Antibody (PE (Phycoerythrin)) [clone: UC10-4F10-11]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 50-1522-U100
  • 50-1522-U025
  • Antigen symbol:
  • CTLA4
  • CTLA4
  • Antigen name:
  • Cytotoxic T-lymphocyte-Associated Protein 4
  • Cytotoxic T-lymphocyte-Associated Protein 4
  • Antigen synonyms:
  • IDDM12|GRD4|GSE|CD|CTLA-4|ALPS5|cytotoxic T-lymphocyte associated protein 4|CD152|CELIAC3
  • IDDM12|GRD4|GSE|CD|CTLA-4|ALPS5|cytotoxic T-lymphocyte associated protein 4|CD152|CELIAC3
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PE (Phycoerythrin)
  • PE (Phycoerythrin)
  • Clone:
  • UC10-4F10-11
  • UC10-4F10-11
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Armenian hamster
  • Armenian hamster
  • Gene ID:
  • 12477
  • 12477
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Flow cytometry:
  • Yes
  • Yes
  • Neutralisation:
  • Yes
  • Yes
  • Cross adsorption:
  • none
  • none
  • Storage temperature:
  • Store undiluted at 2...8 °C, protect from light, do not freeze
  • Store undiluted at 2...8 °C, protect from light, do not freeze
  • Shipping temperature:
  • Ice
  • Ice
  • Tested applications:
  • Flow Cytometry
  • Flow Cytometry

Specifications

Related Information

Frequently Bought Together