The OKT3 antibody is specific for human CD3e, also known as CD3 epsilon, a 20 kDa subunit of the T cell receptor complex along with CD3 gamma and CD3 delta. These integral membrane protein chains assemble with additional chains of the T cell receptor (TCR), as well as CD3 zeta chain, to form the T cell receptor – CD3 complex. Together with co-receptors CD4 or CD8, the complex serves to recognize antigens bound to MHC molecules on antigen-presenting cells. These interactions promote T cell receptor signaling (T cell activation), inducing a number of cellular responses including proliferation, differentiation, production of cytokines or activation-induced cell death. CD3 is differentially expressed during thymocyte-to-T cell development and on all mature T cells.
The OKT3 antibody is a widely used phenotypic marker for human T cells. In addition, as the CD3e subunit of the TCR complex contains intracellular signaling domains, binding of OKT3 antibody to CD3e can induce cell activation. A recent publication of the crystal structure of a CD3e-mitogenic antibody complex provides further insight as to the action of commonly used agonist antibodies, as well as specific epitope-binding data for the widely used human CD3 antibodies OKT3 and UCHT1 (Fernandes, R.A. et al. 2012. J. Biol. Chem. 287: 13324-13335). OKT3 has also been shown to be cross-reactive with Chimpanzee CD3 and has been used for in vitro activation of T cells in this species. Please choose the appropriate format for each application.
Type: Primary
Antigen: CD3E (CD3e molecule, epsilon (CD3-TCR complex))
Clonality: monoclonal
Clone: OKT3
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
- Catalog No:
- 35-0037-T025
- 35-0037-T100
- Antigen symbol:
- CD3E
- CD3E
- Antigen name:
- CD3 Molecule Epsilon
- CD3 Molecule Epsilon
- Antigen synonyms:
- CD3e antigen|TCRE|epsilon subunit of T3|epsilon polypeptide TiT3 complex|T3E|FLJ18683|CD3-epsilon|T-cell surface glycoprotein CD3 epsilon chain|T-cell antigen receptor complex|T-cell surface antigen T3/Leu-4 epsilon chain
- CD3e antigen|TCRE|epsilon subunit of T3|epsilon polypeptide TiT3 complex|T3E|FLJ18683|CD3-epsilon|T-cell surface glycoprotein CD3 epsilon chain|T-cell antigen receptor complex|T-cell surface antigen T3/Leu-4 epsilon chain
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Clone:
- OKT3
- OKT3
- Reactivity:
- Chimpanzee, Human
- Chimpanzee, Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 916
- 916
- Isotype:
- IgG2a kappa
- IgG2a kappa
- Flow cytometry:
- Yes
- Yes
- Neutralisation:
- Yes
- Yes
- Cross adsorption:
- none
- none
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
- Tested applications:
- Flow cytometry
- Flow cytometry