Order Entry
Austria
ContactUsLinkComponent
Anti-IL2 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: JES6-5H4]
Anti-IL2 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: JES6-5H4]
Supplier:  Tonbo Biosciences
Certificates
undefined
Anti-IL2 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: JES6-5H4]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 20-7021-U100
  • 20-7021-U025
  • Antigen symbol:
  • IL2
  • IL2
  • Antigen name:
  • Interleukin 2
  • Interleukin 2
  • Antigen synonyms:
  • POIL2|TCGF|IL-2|lymphokine
  • POIL2|TCGF|IL-2|lymphokine
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • APC (Allophycocyanin)
  • APC (Allophycocyanin)
  • Clone:
  • JES6-5H4
  • JES6-5H4
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 16183
  • 16183
  • Isotype:
  • IgG2b kappa
  • IgG2b kappa
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Cross adsorption:
  • none
  • none
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information

Frequently Bought Together