Order Entry
Austria
ContactUsLinkComponent
Anti-KLRG1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 2F1]
Anti-KLRG1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 2F1]
Supplier:  Tonbo Biosciences
Certificates
undefined
Anti-KLRG1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 2F1]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 20-5893-U025
  • 20-5893-U100
  • Antigen symbol:
  • KLRG1
  • KLRG1
  • Antigen name:
  • Killer cell lectin-like receptor subfamily G member 1
  • Killer cell lectin-like receptor subfamily G member 1
  • Antigen synonyms:
  • MAFA|MAFA-2F1|2F1|CLEC15A|MAFA-LIKE|MAFA-L
  • MAFA|MAFA-2F1|2F1|CLEC15A|MAFA-LIKE|MAFA-L
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • APC (Allophycocyanin)
  • APC (Allophycocyanin)
  • Clone:
  • 2F1
  • 2F1
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 50928
  • 50928
  • Isotype:
  • IgG
  • IgG
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Neutralisation:
  • Yes
  • Yes
  • Cross adsorption:
  • none
  • none
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice
  • Tested applications:
  • Flow Cytometry
  • Flow Cytometry

Specifications

Related Information

Frequently Bought Together