Order Entry
Austria
ContactUsLinkComponent
Anti-Thy1.2 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 30-H12]
Anti-Thy1.2 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 30-H12]
Supplier:  Tonbo Biosciences
Certificates
undefined
Anti-Thy1.2 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 30-H12]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 20-0903-U025
  • 20-0903-U100
  • Antigen symbol:
  • THY1
  • THY1
  • Antigen name:
  • Thy1 Cell Surface Antigen
  • Thy1 Cell Surface Antigen
  • Antigen synonyms:
  • Thy-1 T-cell antigen|Thy-1 cell surface antigen|Thy-1 membrane glycoprotein|THY1|FLJ33325
  • Thy-1 T-cell antigen|Thy-1 cell surface antigen|Thy-1 membrane glycoprotein|THY1|FLJ33325
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • APC (Allophycocyanin)
  • APC (Allophycocyanin)
  • Clone:
  • 30-H12
  • 30-H12
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 21838
  • 21838
  • Isotype:
  • IgG2b
  • IgG2b
  • Flow cytometry:
  • Yes
  • Yes
  • Cross adsorption:
  • none
  • none
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information

Frequently Bought Together