Order Entry
Austria
ContactUsLinkComponent
Anti-ITPKA Rabbit Polyclonal Antibody
Anti-ITPKA Rabbit Polyclonal Antibody
Catalog # ABGEAP8166A
Supplier:  Abgent
undefined
Anti-ITPKA Rabbit Polyclonal Antibody
Catalog # ABGEAP8166A
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Inositol-trisphosphate 3-kinase A
  • Antigen symbol:
    ITPKA
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    N-Terminal
  • Cross adsorption:
    No
  • Gene ID:
    3178
  • Antigen synonyms:
    IP3 3-kinase A|Inositol-trisphosphate 3-kinase A|Inositol 1,4,5-trisphosphate 3-kinase A|IP3K A
  • Storage buffer:
    PBS with 0,09% (W/V) sodium azide, pH 7,4
  • Sequence:
    YGGSGSYDSYNNGGC
  • Storage temperature:
    Cool
  • Concentration:
    0,5 mg/ml
  • Shipping temperature:
    Ice
  • Immunogen:
    This HNRNPA1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 266-294 amino acids from the C-terminal region of human HNRNPA1.
  • Purification:
    Protein A & Peptide affinity purification
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: ITPKA (inositol-trisphosphate 3-kinase A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human