Order Entry
Austria
ContactUsLinkComponent
Anti-PHLPP1 Rabbit Polyclonal Antibody
Anti-PHLPP1 Rabbit Polyclonal Antibody
Catalog # ABGEAP7842A
Supplier:  Abgent
undefined
Anti-PHLPP1 Rabbit Polyclonal Antibody
Catalog # ABGEAP7842A
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    PH Domain And leucine rich Repeat Protein phosphatase 1
  • Antigen symbol:
    PHLPP1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    N-terminal
  • Antigen synonyms:
    Suprachiasmatic nucleus circadian oscillatory protein|hSCOP|KIAA0606|PH domain leucine-rich repeat protein phosphatase|SCOP|PHLPP1|PLEKHE1|PHLPP|PH domain leucine-rich repeat-containing protein phosphatase|EC 3.1.3.16|Pleckstrin homology domain-containing family E protein 1
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: PHLPP1 (PH domain and leucine rich repeat protein phosphatase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human