Anti-BCKDHA Rabbit Polyclonal Antibody
Catalog # ABGEAP6830B
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Branched Chain Keto Acid Dehydrogenase E1, Alpha Polypeptide
- Antigen symbol:BCKDHA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Antigen synonyms:BCKDE1A|MSU|OVD1A|MSUD1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: BCKDHA (branched chain keto acid dehydrogenase E1, alpha polypeptide)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human