Anti-BUB1A Rabbit Polyclonal Antibody
Catalog # ABGEAP5438A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:BUB1 mitotic checkpoint serine/threonine kinase
- Antigen symbol:BUB1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Antigen synonyms:hBUB1|BUB1L|BUB1A
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: BUB1A
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human