Anti-MPP4 Rabbit Polyclonal Antibody
Catalog # AVIVARP65025_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Membrane Protein, Palmitoylated 4 (MAGUK p55 subfamily member 4)
- Antigen symbol:MPP4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:58538
- Antigen synonyms:DLG6|ALS2CR5
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: MPP4 (membrane protein, palmitoylated 4 (MAGUK p55 subfamily member 4))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human