Anti-LONP2 Rabbit Polyclonal Antibody
Catalog # AVIVARP60626_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:lon peptidase 2, peroxisomal
- Antigen symbol:LONP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:83752
- Antigen synonyms:LONPL|LONP
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: LONP2 (lon peptidase 2, peroxisomal)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: