Order Entry
Austria
ContactUsLinkComponent
Anti-SLC25A19 Rabbit Polyclonal Antibody
Anti-SLC25A19 Rabbit Polyclonal Antibody
  AVIVARP44023_P050
Anti-SLC25A19 Rabbit Polyclonal Antibody
  AVIVARP44023_P050

 

  • Antibody type:
    Primary
  • Antigen name:
    solute carrier family 25 (mitochondrial thiamine pyrophosphate carrier), member 19
  • Antigen symbol:
    Solute carrier family 25, member 19
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Western blot:
    Yes
  • Epitope:
    middle
  • Cross adsorption:
    none
  • Gene ID:
    60386
  • Antigen synonyms:
    THMD3|MUP1|THMD4|DNC|TPC|MCPHA
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    50 µG

 

 

Type: Primary
Antigen: SLC25A19 (solute carrier family 25 (mitochondrial thiamine pyrophosphate carrier), member 19)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: