Anti-SLC30A1 Rabbit Polyclonal Antibody
Catalog # AVIVARP44019_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:solute carrier family 30 (zinc transporter), member 1
- Antigen symbol:Solute carrier family 30, member 1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:7779
- Antigen synonyms:ZNT1|ZRC1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: SLC30A1 (solute carrier family 30 (zinc transporter), member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: