Order Entry
Austria
ContactUsLinkComponent
 
Anti-ADH4 Rabbit Polyclonal Antibody
Catalog # AVIVARP43573_T100
Anti-ADH4 Rabbit Polyclonal Antibody
Catalog # AVIVARP43573_T100

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    alcohol dehydrogenase 4 (class II), pi polypeptide
  • Antigen symbol:
    ADH4
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
  • Reactivity:
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    middle
  • Cross adsorption:
    none
  • Gene ID:
    127
  • Antigen synonyms:
    ADH-2|HEL-S-4
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: ADH4 (alcohol dehydrogenase 4 (class II), pi polypeptide)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: